Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012765.4 | NP_001012783.3 | 12 - 13 | 2904 | 81 |
GenBank mRNA | AK147590.1 | - | - | 3007 | 81 |
AK147649.1 | - | - | 3664 | 81 | |
AK160942.1 | - | 12 - 13 | 2904 | 81 | |
BC035550.1 | - | - | 1042 | 81 | |
BC090846.1 | - | 12 - 13 | 2639 | 81 |