Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028803.4 | NP_083079.1 | 12 - 13 | 1821 | 60 |
GenBank mRNA | AK009815.1 | - | 12 - 13 | 1637 | 60 |
AK050365.1 | - | 12 - 13 | 1708 | 60 | |
AK050423.1 | - | 12 - 13 | 1707 | 60 | |
AK148186.1 | - | 6 - 7 | 749 | 60 | |
BC017541.1 | - | 12 - 13 | 1682 | 60 |