Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028803.4 | NP_083079.1 | 14 - 15 | 2138 | 58 |
GenBank mRNA | AK009815.1 | - | 14 - 15 | 1954 | 58 |
AK050365.1 | - | 14 - 15 | 2025 | 58 | |
AK210800.1 | - | 1 - 2 | 126 | 58 | |
BC017541.1 | - | 14 - 15 | 1999 | 58 |