Assay Design
Gene Transcripts
- Gene Symbol
- Crybg3
- Entrez Gene ID
-
224273
- Gene Name
- beta-gamma crystallin domain containing 3
- Gene Aliases
-
5031404N19, AU022790
- Chromosome Location
-
Chr.16: 59490775 - 59601399 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_174848.3 |
NP_777273.2 |
4 - 5 |
1952 |
64 |
GenBank mRNA |
BC043118.1 |
- |
4 - 5 |
1952 |
64 |
View Full Product Details