Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011782.2 | NP_035912.2 | 7 - 8 | 3087 | 133 |
GenBank mRNA | AF140673.1 | - | 7 - 8 | 2246 | 133 |
AK044746.1 | - | 7 - 8 | 1309 | 133 | |
AK082475.1 | - | 7 - 8 | 1309 | 133 | |
BC138619.1 | - | 7 - 8 | 2271 | 133 | |
BC138620.1 | - | 7 - 8 | 2271 | 133 |