Assay Design
Gene Transcripts
- Gene Symbol
- Tmem30c
- Entrez Gene ID
-
71027
- Gene Name
- transmembrane protein 30C
- Gene Aliases
-
4933401B01Rik, 4933409A18Rik, Cdc50c
- Chromosome Location
-
Chr.16: 57266138 - 57301893 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_027651.1 |
NP_081927.1 |
1 - 2 |
62 |
68 |
GenBank mRNA |
AK016597.1 |
- |
1 - 2 |
76 |
68 |
View Full Product Details