Assay Design



Gene Transcripts

Gene Symbol
Pigx
Entrez Gene ID
72084
Gene Name
phosphatidylinositol glycan anchor biosynthesis, class X
Gene Aliases
2010319C14Rik, PIG-X
Chromosome Location
Chr.16: 32084416 - 32099727 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024464.3 NP_077784.2 1 - 2 264 134
GenBank mRNA AK008583.2 - 1 - 2 266 134
BY708475.1 - 1 - 2 134

View Full Product Details