Assay Design
Gene Transcripts
- Gene Symbol
- Thap6
- Entrez Gene ID
-
381650
- Gene Name
- THAP domain containing 6
- Gene Aliases
-
4930534K21Rik, AI448650
- Chromosome Location
-
Chr.5: 91962882 - 91972137 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK015967.1 |
- |
1 - 2 |
584 |
82 |
View Full Product Details