Assay Design
Gene Transcripts
- Gene Symbol
- Cxcl10
- Entrez Gene ID
-
15945
- Gene Name
- chemokine (C-X-C motif) ligand 10
- Gene Aliases
-
C7, CRG-2, INP10, IP-10, IP10, Ifi10, Scyb10, gIP-10, mob-1
- Chromosome Location
-
Chr.5: 92346638 - 92348889 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AF227743.1 |
- |
2 - 3 |
222 |
151 |
AJ243095.1 |
- |
1 - 2 |
73 |
151 |
AK146144.1 |
- |
2 - 3 |
259 |
151 |
M86829.1 |
- |
2 - 3 |
251 |
151 |
View Full Product Details