Assay Design



Gene Transcripts

Gene Symbol
Ptgr1
Entrez Gene ID
67103
Gene Name
prostaglandin reductase 1
Gene Aliases
2510002C21Rik, Ltb4dh, Zadh3
Chromosome Location
Chr.4: 58965590 - 58987131 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025968.3 NP_080244.1 10 - 10 1644 99
GenBank mRNA AK035425.1 - 10 - 10 1645 99
AK134440.1 - 10 - 10 1676 99

View Full Product Details