Assay Design



Gene Transcripts

Gene Symbol
Thap6
Entrez Gene ID
381650
Gene Name
THAP domain containing 6
Gene Aliases
4930534K21Rik, AI448650
Chromosome Location
Chr.5: 91962882 - 91972137 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_028429.1 4 - 5 1082 68
GenBank mRNA AK039179.1 - 3 - 4 958 68
AK080650.1 - 3 - 4 957 68
BC089377.1 - 4 - 5 631 68

View Full Product Details