Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001098476.2 | NP_001091946.1 | 7 - 8 | 976 | 94 |
NM_178700.5 | NP_848815.2 | 7 - 8 | 1304 | 94 | |
GenBank mRNA | AK044889.1 | - | 7 - 8 | 1305 | 94 |
AK045986.1 | - | 7 - 8 | 1258 | 94 | |
AK077782.1 | - | 7 - 8 | 1305 | 94 | |
AK089014.1 | - | 6 - 7 | 840 | 94 | |
AK168385.1 | - | 7 - 8 | 948 | 94 | |
AK168459.1 | - | 7 - 8 | 948 | 94 |