Assay Design



Gene Transcripts

Gene Symbol
Nktr
Entrez Gene ID
18087
Gene Name
natural killer tumor recognition sequence
Gene Aliases
5330401F18Rik, D9Wsu172e
Chromosome Location
Chr.9: 121718181 - 121756842 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010918.2 NP_035048.3 5 - 6 421 86
GenBank mRNA AK035582.1 - 4 - 5 322 86
AK144214.1 - 5 - 6 332 86
L04289.2 - 5 - 6 426 86

View Full Product Details