Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001172136.1 | NP_001165607.1 | 6 - 7 | 743 | 123 |
NM_172456.3 | NP_766044.1 | 5 - 6 | 708 | 123 | |
GenBank mRNA | AK028887.1 | - | 5 - 6 | 672 | 123 |
AK034654.1 | - | 6 - 7 | 705 | 123 | |
AK051300.1 | - | 6 - 7 | 707 | 123 | |
AK148426.1 | - | 5 - 6 | 689 | 123 | |
AK162566.1 | - | 5 - 6 | 671 | 123 | |
AK163555.1 | - | 5 - 6 | 672 | 123 | |
BC052171.1 | - | 4 - 5 | 514 | 123 | |
BC059920.1 | - | 6 - 7 | 686 | 123 | |
BC064730.1 | - | 6 - 7 | 758 | 123 | |
BC094589.1 | - | 3 - 4 | 3194 | 123 | |
BC119024.1 | - | 5 - 6 | 672 | 123 | |
BC144729.1 | - | 6 - 7 | 707 | 123 |