Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001122896.2 | NP_001116368.1 | 4 - 5 | 554 | 65 |
NM_001122897.2 | NP_001116369.1 | 5 - 6 | 791 | 65 | |
NM_001285478.1 | NP_001272407.1 | 5 - 6 | 811 | 65 | |
NM_001285479.1 | NP_001272408.1 | 5 - 6 | 752 | 65 | |
NM_001285480.1 | NP_001272409.1 | 4 - 5 | 515 | 65 | |
NM_001285481.1 | NP_001272410.1 | 5 - 6 | 713 | 65 | |
NM_010470.3 | NP_034600.2 | 5 - 6 | 752 | 65 | |
GenBank mRNA | AK004938.1 | - | 5 - 6 | 729 | 65 |
AK028181.1 | - | 5 - 6 | 730 | 65 | |
AK043260.1 | - | 4 - 5 | 528 | 65 | |
AK050234.1 | - | 5 - 6 | 731 | 65 | |
AK075725.1 | - | 4 - 5 | 493 | 65 | |
AK150784.1 | - | 5 - 6 | 797 | 65 | |
AK161041.1 | - | 4 - 5 | 522 | 65 | |
AK166161.1 | - | 5 - 6 | 699 | 65 | |
AK169740.1 | - | 5 - 6 | 658 | 65 | |
BC020024.1 | - | 5 - 6 | 702 | 65 | |
BC107687.1 | - | 5 - 6 | 687 | 65 | |
BU612149.1 | - | 4 - 5 | 555 | 65 | |
X99642.1 | - | 3 - 4 | 395 | 65 |