Assay Design



Gene Transcripts

Gene Symbol
Fbxo8
Entrez Gene ID
50753
Gene Name
F-box protein 8
Gene Aliases
Fbx8
Chromosome Location
Chr.8: 56551134 - 56593939 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015791.3 NP_056606.2 2 - 3 582 70
GenBank mRNA AF176527.1 - 2 - 3 589 70
AK002538.1 - 2 - 3 593 70
AK012401.1 - 2 - 3 600 70
AK029989.1 - 2 - 3 583 70
AK077595.1 - 2 - 3 604 70
AK078111.1 - 2 - 3 602 70
AK147748.1 - 2 - 3 599 70
AK160145.1 - 2 - 3 563 70
AK162592.1 - 2 - 3 599 70
BC011100.1 - 2 - 3 603 70
BC034854.1 - 2 - 3 514 70

View Full Product Details