Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081355.3 | NP_001074824.3 | 2 - 3 | 166 | 108 |
NM_001256380.1 | NP_001243309.1 | 2 - 3 | 166 | 108 | |
GenBank mRNA | AK087820.1 | - | 2 - 3 | 167 | 108 |
BB556664.2 | - | 2 - 3 | 176 | 108 | |
BC151062.1 | - | 1 - 2 | 79 | 108 |