Assay Design



Gene Transcripts

Gene Symbol
Prdm2
Entrez Gene ID
110593
Gene Name
PR domain containing 2, with ZNF domain
Gene Aliases
4833427P12Rik, E330024L24Rik, Gm1033, Gm19732, KMT8, Riz, Riz1, Znfpr1c1
Chromosome Location
Chr.4: 143107391 - 143212737 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081355.3 NP_001074824.3 5 - 6 543 94
NM_001256380.1 NP_001243309.1 5 - 6 543 94
GenBank mRNA AK087820.1 - 5 - 6 517 94
BB556664.2 - 5 - 6 526 94
BC151062.1 - 4 - 5 456 94

View Full Product Details