Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001007570.2 | NP_001007571.1 | 5 - 6 | 465 | 92 |
GenBank mRNA | BC022213.1 | - | 5 - 6 | 455 | 92 |
BC025937.1 | - | 5 - 6 | 440 | 92 | |
BC036140.1 | - | 5 - 6 | 444 | 92 |