Assay Design



Gene Transcripts

Gene Symbol
Slc25a42
Entrez Gene ID
73095
Gene Name
solute carrier family 25, member 42
Gene Aliases
2900084M01Rik
Chromosome Location
Chr.8: 70184340 - 70212281 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007570.2 NP_001007571.1 6 - 7 587 64
GenBank mRNA AK143301.1 - 5 - 6 1049 64
AK165793.1 - 5 - 6 1050 64
BC022213.1 - 6 - 7 577 64
BC025937.1 - 6 - 7 562 64
BC036140.1 - 6 - 7 566 64

View Full Product Details