Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001111066.1 | NP_001104536.1 | 7 - 8 | 1110 | 66 |
NM_001199631.1 | NP_001186560.1 | 7 - 8 | 1206 | 66 | |
NM_010223.2 | NP_034353.2 | 7 - 8 | 1107 | 66 | |
GenBank mRNA | AB154247.1 | - | 6 - 7 | 846 | 66 |
AF030635.1 | - | 6 - 7 | 877 | 66 | |
AK088739.1 | - | 7 - 8 | 1045 | 66 | |
AK146122.1 | - | 7 - 8 | 1105 | 66 | |
AK152198.1 | - | 7 - 8 | 1106 | 66 | |
AK165281.1 | - | 7 - 8 | 1103 | 66 | |
AK167663.1 | - | 7 - 8 | 1093 | 66 | |
AK189147.1 | - | 1 - 2 | 88 | 66 | |
AY225340.1 | - | 6 - 7 | 990 | 66 | |
AY278608.1 | - | 7 - 8 | 1066 | 66 | |
BC003739.1 | - | 6 - 7 | 1026 | 66 | |
BC027808.1 | - | 7 - 8 | 1060 | 66 |