Assay Design



Gene Transcripts

Gene Symbol
Fkbp8
Entrez Gene ID
14232
Gene Name
FK506 binding protein 8
Gene Aliases
38kDa, FKBP-38, FKBP-8, Fkbp38
Chromosome Location
Chr.8: 70527743 - 70535328 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199631.1 NP_001186560.1 1 - 2 191 75

View Full Product Details