Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166463.1 | NP_001159935.1 | 6 - 7 | 734 | 54 |
NM_001166464.1 | NP_001159936.1 | 7 - 8 | 743 | 54 | |
NM_001166465.1 | NP_001159937.1 | 6 - 7 | 734 | 54 | |
NM_001166466.1 | NP_001159938.1 | 7 - 8 | 743 | 54 | |
NM_009262.3 | NP_033288.2 | 7 - 8 | 743 | 54 | |
GenBank mRNA | AK036189.1 | - | 7 - 8 | 744 | 54 |
AK039058.1 | - | 6 - 7 | 716 | 54 | |
AK049019.1 | - | 6 - 7 | 702 | 54 | |
AK051191.1 | - | 6 - 7 | 694 | 54 | |
BC075625.1 | - | 7 - 8 | 686 | 54 | |
BC158020.1 | - | 7 - 8 | 650 | 54 | |
X92864.1 | - | 7 - 8 | 735 | 54 |