Assay Design



Gene Transcripts

Gene Symbol
Pomgnt2
Entrez Gene ID
215494
Gene Name
protein O-linked mannose beta 1,4-N-acetylglucosaminyltransferase 2
Gene Aliases
Ago61, C85492, Gtdc2
Chromosome Location
Chr.9: 121981606 - 121996026 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289559.1 NP_001276488.1 2 - 3 210 70
GenBank mRNA AK054403.1 - 2 - 3 198 70

View Full Product Details