Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012007.1 | NP_036137.1 | 4 - 5 | 472 | 70 |
GenBank mRNA | AF136272.1 | - | 4 - 5 | 472 | 70 |
AF158182.1 | - | 3 - 4 | 412 | 70 | |
AK005526.1 | - | 4 - 5 | 479 | 70 | |
AK131661.1 | - | 4 - 5 | 479 | 70 | |
BC103769.1 | - | 4 - 5 | 468 | 70 |