Assay Design



Gene Transcripts

Gene Symbol
Kng2
Entrez Gene ID
385643
Gene Name
kininogen 2
Gene Aliases
-
Chromosome Location
Chr.16: 22985852 - 23029101 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102410.1 NP_001095880.1 10 - 11 1401 68
GenBank mRNA AY462057.1 - 10 - 11 1317 68
BC108934.1 - 10 - 11 1259 68
BC139720.1 - 10 - 11 1309 68

View Full Product Details