Assay Design
Gene Transcripts
- Gene Symbol
- Faf2
- Entrez Gene ID
-
76577
- Gene Name
- Fas associated factor family member 2
- Gene Aliases
-
2210404D11Rik, AI462440, Ubxd8, mKIAA0887
- Chromosome Location
-
Chr.13: 54621775 - 54664063 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC048767.1 |
- |
2 - 3 |
143 |
76 |
View Full Product Details