Assay Design
Gene Transcripts
- Gene Symbol
- Hp1bp3
- Entrez Gene ID
-
15441
- Gene Name
- heterochromatin protein 1, binding protein 3
- Gene Aliases
-
Hp1bp74
- Chromosome Location
-
Chr.4: 138216582 - 138244683 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BY130345.1 |
- |
1 - 2 |
164 |
59 |
CN670198.1 |
- |
1 - 2 |
168 |
59 |
View Full Product Details