Assay Design



Gene Transcripts

Gene Symbol
Melk
Entrez Gene ID
17279
Gene Name
maternal embryonic leucine zipper kinase
Gene Aliases
AI327312, MPK38, mKIAA0175
Chromosome Location
Chr.4: 44300876 - 44364675 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010790.2 NP_034920.2 2 - 3 254 101
GenBank mRNA AK010498.1 - 2 - 3 221 101
AK011932.1 - 2 - 3 254 101
AK129076.1 - 2 - 3 207 101
AK137000.1 - 2 - 3 279 101
AK145021.1 - 2 - 3 189 101
AK145316.1 - 2 - 3 163 101
AK161029.1 - 2 - 3 263 101
AK164138.1 - 2 - 3 221 101
AK167457.1 - 2 - 3 178 101
AK208560.1 - 1 - 2 112 102
BC020416.1 - 1 - 2 89 101
BC085276.1 - 2 - 3 287 101
L76158.1 - 2 - 3 185 101
X95351.1 - 2 - 3 192 101

View Full Product Details