Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289915.1 | NP_001276844.1 | 2 - 3 | 260 | 66 |
NM_009856.3 | NP_033986.1 | 2 - 3 | 260 | 66 | |
GenBank mRNA | AF001036.1 | - | 2 - 3 | 158 | 66 |
AJ245551.1 | - | 2 - 3 | 171 | 66 | |
AK075888.1 | - | 2 - 3 | 190 | 66 | |
AK139410.1 | - | 2 - 3 | 188 | 66 | |
AK154862.1 | - | 2 - 3 | 188 | 66 | |
AK156386.1 | - | 2 - 3 | 189 | 66 | |
AK171716.1 | - | 2 - 3 | 189 | 66 | |
AK172122.1 | - | 2 - 3 | 188 | 66 | |
AK172396.1 | - | 2 - 3 | 176 | 66 | |
AK172449.1 | - | 2 - 3 | 189 | 66 | |
AK172466.1 | - | 2 - 3 | 183 | 66 | |
AK172515.1 | - | 1 - 2 | 283 | 66 | |
BB840485.1 | - | 2 - 3 | 260 | 66 | |
BC072615.1 | - | 2 - 3 | 176 | 66 | |
BC107344.1 | - | 2 - 3 | 171 | 66 | |
BC107345.1 | - | 2 - 3 | 171 | 66 | |
BY336480.1 | - | 2 - 3 | 266 | 66 |