Assay Design



Gene Transcripts

Gene Symbol
Kazn
Entrez Gene ID
71529
Gene Name
kazrin, periplakin interacting protein
Gene Aliases
2310007B04Rik, 9030409G11Rik, AI256607, AI666764, AW061099, Kaz, Kazrin
Chromosome Location
Chr.4: 142102390 - 142239401 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144531.3 NP_653114.3 13 - 14 2333 95
GenBank mRNA AK032460.1 - 11 - 12 1572 95
AK173090.1 - 13 - 14 2333 95

View Full Product Details