Assay Design
Gene Transcripts
- Gene Symbol
- Nktr
- Entrez Gene ID
-
18087
- Gene Name
- natural killer tumor recognition sequence
- Gene Aliases
-
5330401F18Rik, D9Wsu172e
- Chromosome Location
-
Chr.9: 121718181 - 121756842 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_010918.2 |
NP_035048.3 |
16 - 17 |
4418 |
82 |
GenBank mRNA |
AK144214.1 |
- |
16 - 17 |
4329 |
82 |
L04289.2 |
- |
16 - 17 |
4423 |
82 |
View Full Product Details