Assay Design



Gene Transcripts

Gene Symbol
Fktn
Entrez Gene ID
246179
Gene Name
fukutin
Gene Aliases
D830030O17Rik, Fcmd
Chromosome Location
Chr.4: 53714107 - 53765785 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_139309.4 NP_647470.1 4 - 5 762 137
GenBank mRNA AB077383.1 - 5 - 6 946 137
AJ511807.1 - 5 - 6 782 137
AK052357.1 - 6 - 7 920 137
AK085931.1 - 4 - 5 763 137
AK142744.1 - - 922 137
AK144743.1 - 5 - 6 791 137
BC017538.1 - 5 - 6 778 137

View Full Product Details