Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163776.1 | NP_001157248.1 | 10 - 11 | 1241 | 74 |
NM_080727.2 | NP_542765.2 | 9 - 10 | 1101 | 74 | |
GenBank mRNA | AJ300738.1 | - | 9 - 10 | 1101 | 74 |
AK158203.1 | - | 10 - 11 | 1244 | 74 | |
BC112375.1 | - | 9 - 10 | 1098 | 74 |