Assay Design



Gene Transcripts

Gene Symbol
Foxp3
Entrez Gene ID
20371
Gene Name
forkhead box P3
Gene Aliases
JM2, scurfin, sf
Chromosome Location
Chr.X: 7579676 - 7595243 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199347.1 NP_001186276.1 7 - 8 881 72
NM_001199348.1 NP_001186277.1 6 - 7 739 72
NM_054039.2 NP_473380.1 6 - 7 814 72
GenBank mRNA AF277991.1 - 6 - 7 739 72
AF277992.1 - 6 - 7 797 72
AK157447.1 - 7 - 8 882 72
AY357712.1 - 5 - 6 597 72
AY357713.1 - 5 - 6 597 72
BC132333.1 - 5 - 6 585 72
BC132335.1 - 5 - 6 585 72
DQ387959.1 - 7 - 8 881 72
KF471319.1 - 5 - 6 581 72

View Full Product Details