Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015791.3 | NP_056606.2 | 3 - 4 | 705 | 78 |
GenBank mRNA | AF176527.1 | - | 3 - 4 | 712 | 78 |
AK002538.1 | - | 3 - 4 | 716 | 78 | |
AK029989.1 | - | 3 - 4 | 706 | 78 | |
AK077595.1 | - | 3 - 4 | 727 | 78 | |
AK078111.1 | - | 3 - 4 | 725 | 78 | |
AK147748.1 | - | 3 - 4 | 722 | 78 | |
BC034854.1 | - | 3 - 4 | 637 | 78 |