Assay Design



Gene Transcripts

Gene Symbol
Fam47e
Entrez Gene ID
384198
Gene Name
family with sequence similarity 47, member E
Gene Aliases
C730043O17, Gm1381
Chromosome Location
Chr.5: 92554951 - 92591285 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033478.2 NP_001028650.2 3 - 4 70
GenBank mRNA AK144479.1 - 3 - 4 70

View Full Product Details