Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025495.3 | NP_079771.1 | 7 - 8 | 774 | 94 |
GenBank mRNA | AK006234.1 | - | 7 - 8 | 811 | 94 |
AK012863.1 | - | 7 - 8 | 778 | 94 | |
BC094280.1 | - | 7 - 8 | 827 | 94 | |
BC147207.1 | - | 7 - 8 | 754 | 94 | |
BC147208.1 | - | 7 - 8 | 754 | 94 |