Assay Design
Gene Transcripts
- Gene Symbol
- Phactr1
- Entrez Gene ID
-
218194
- Gene Name
- phosphatase and actin regulator 1
- Gene Aliases
-
9630030F18Rik, Rpel1
- Chromosome Location
-
Chr.13: 42680571 - 43138526 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK047654.1 |
- |
2 - 3 |
281 |
83 |
AY993932.1 |
- |
2 - 3 |
267 |
83 |
View Full Product Details