Assay Design
Gene Transcripts
- Gene Symbol
- Dsp
- Entrez Gene ID
-
109620
- Gene Name
- desmoplakin
- Gene Aliases
-
2300002E22Rik, 5730453H04Rik, AA407887, AA407888, AW109828, DP, rul
- Chromosome Location
-
Chr.13: 38151084 - 38198577 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_023842.2 |
NP_076331.2 |
8 - 9 |
1378 |
66 |
View Full Product Details