Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001146153.1 | NP_001139625.1 | 1 - 2 | 100 | 81 |
NM_011984.2 | NP_036114.2 | 1 - 2 | 100 | 81 | |
GenBank mRNA | BC005773.1 | - | 1 - 2 | 49 | 81 |
BC095961.1 | - | 1 - 2 | 55 | 81 | |
CX208892.1 | - | 1 - 2 | 79 | 81 |