Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011606.2 | NP_035736.2 | 1 - 2 | 205 | 98 |
GenBank mRNA | BC035043.1 | - | 1 - 2 | 157 | 98 |
BY114395.1 | - | 1 - 2 | 205 | 98 | |
U08595.1 | - | 1 - 2 | 188 | 98 | |
X79199.1 | - | 1 - 2 | 180 | 98 |