Assay Design



Gene Transcripts

Gene Symbol
Cyp4f13
Entrez Gene ID
170716
Gene Name
cytochrome P450, family 4, subfamily f, polypeptide 13
Gene Aliases
0610030I10Rik, Cypf13
Chromosome Location
Chr.17: 32924688 - 32947415 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130882.1 NP_570952.1 5 - 6 576 107
GenBank mRNA AF233643.1 - 5 - 6 576 107
BC003954.1 - 5 - 6 585 107

View Full Product Details