Assay Design



Gene Transcripts

Gene Symbol
Gabbr2
Entrez Gene ID
242425
Gene Name
gamma-aminobutyric acid (GABA) B receptor, 2
Gene Aliases
GABABR2, Gb2, Gm425, Gpr51
Chromosome Location
Chr.4: 46663898 - 46991714 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081141.1 NP_001074610.1 17 - 18 2828 76
GenBank mRNA AK132744.1 - 5 - 6 1322 76

View Full Product Details