Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008673.1 | NP_032699.1 | 1 - 1 | 1053 | 160 |
GenBank mRNA | BC119181.1 | - | 1 - 1 | 858 | 160 |
BC137747.1 | - | 1 - 1 | 858 | 160 | |
U35885.1 | - | 1 - 1 | 1053 | 160 |