Assay Design



Gene Transcripts

Gene Symbol
Umodl1
Entrez Gene ID
52020
Gene Name
uromodulin-like 1
Gene Aliases
D17Ertd488e
Chromosome Location
Chr.17: 30954640 - 31010710 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177465.4 NP_803416.2 19 - 20 3851 60
GenBank mRNA AK040446.1 - 11 - 12 2352 60
AY771616.1 - 20 - 21 3768 60
AY771617.1 - 19 - 20 3855 60
AY771618.1 - 19 - 20 3510 60
BC051427.1 - 19 - 20 3506 60
BC132381.1 - 6 - 7 1040 60

View Full Product Details