Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008176.3 | NP_032202.1 | 1 - 2 | 136 | 87 |
GenBank mRNA | BC037997.1 | - | 1 - 2 | 103 | 87 |
BC132502.1 | - | 1 - 2 | 95 | 87 | |
BC132504.1 | - | 1 - 2 | 95 | 87 | |
BY335927.1 | - | 1 - 2 | 143 | 87 | |
J04596.1 | - | 1 - 2 | 71 | 87 |