Assay Design
Gene Transcripts
- Gene Symbol
- Mthfd2l
- Entrez Gene ID
-
665563
- Gene Name
- methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 2-like
- Gene Aliases
-
1110019K23Rik, C630010D07Rik
- Chromosome Location
-
Chr.5: 90931196 - 91021368 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_026788.1 |
NP_081064.1 |
6 - 7 |
900 |
69 |
GenBank mRNA |
BU610697.1 |
- |
5 - 6 |
581 |
69 |
View Full Product Details