Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001101430.2 | NP_001094900.1 | 1 - 2 | 284 | 84 |
GenBank mRNA | AA063946.1 | - | 1 - 2 | 270 | 84 |
AA124813.1 | - | 1 - 2 | 160 | 84 | |
BC070417.1 | - | 1 - 2 | 258 | 84 |