Assay Design
Gene Transcripts
- Gene Symbol
- Padi3
- Entrez Gene ID
-
18601
- Gene Name
- peptidyl arginine deiminase, type III
- Gene Aliases
-
AI507004, Pad3, Pdi3, mKIAA4171
- Chromosome Location
-
Chr.4: 140785369 - 140810648 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_011060.4 |
NP_035190.3 |
7 - 8 |
868 |
69 |
GenBank mRNA |
AK220522.1 |
- |
6 - 7 |
1110 |
69 |
View Full Product Details